Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL18A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030582.3 | NP_085059.2 | 2 | 826 | ||
NM_130444.2 | NP_569711.2 | 2 | 1531 | |||
NM_130445.3 | NP_569712.2 | 3 | 359 | |||
GenBank mRNA | AF018081.1 | - | 2 | 845 | ||
AF018082.1 | - | 3 | 324 | |||
BC063833.1 | - | 3 | 300 | 6181818 | ||
CN389577.1 | - | 3 | 359 |