Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other AGPAT3 SIRNA Assays ›
Gene Symbol:
AGPAT3
Gene Name:
1-acylglycerol-3-phosphate O-acyltransferase 3
Gene Aliases:
LPAAT-GAMMA1, LPAAT3
Chromosome Location:
Chr.21: 43865160 - 43987594 on Build GRCh38
UniGene ID:
Hs.248785
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
43694
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: AGPAT3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001037553.1
NP_001032642.1
3
326
NM_020132.4
NP_064517.1
4
446
XM_005261160.4
XP_005261217.1
4
447
XM_006724029.3
XP_006724092.1
3
375
XM_006724030.3
XP_006724093.1
3
343
XM_006724031.3
XP_006724094.1
3
346
XM_011529660.2
XP_011527962.1
3
185
XM_011529661.2
XP_011527963.1
3
500
XM_011529662.2
XP_011527964.1
5
1150
XM_011529663.2
XP_011527965.1
3
359
XM_011529664.2
XP_011527966.1
3
300
XM_011529665.2
XP_011527967.1
5
801
XM_017028408.1
XP_016883897.1
3
329
XM_017028409.1
XP_016883898.1
3
407
XM_017028410.1
XP_016883899.1
3
412
XM_017028411.1
XP_016883900.1
3
285
XM_017028412.1
XP_016883901.1
4
1378
XM_017028413.1
XP_016883902.1
4
346
GenBank mRNA
AB040138.1
-
3
401
AF156774.1
-
4
407
AF156775.1
-
3
270
AK125804.1
-
1
3063
AK300562.1
-
3
328
AY358704.1
-
495
BC011971.1
-
4
420
4148915
BC040603.1
-
5
1150
5270905
BC063552.1
-
3
716
4800514
BM083518.1
-
3
326
CD672915.1
-
4
446
Back To Top
More Information
Panther Classification:
Molecular Function -
acyltransferase
Gene Ontology Categories:
Function(s)
Process(es)
phosphatidic acid biosynthetic process
phospholipid biosynthetic process
CDP-diacylglycerol biosynthetic process
triglyceride biosynthetic process
1-acylglycerol-3-phosphate O-acyltransferase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-d4wl8:80/10.239.162.132:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0