Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNK17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135111.1 | NP_001128583.1 | 2 | 399 | ||
NM_031460.3 | NP_113648.2 | 2 | 398 | |||
GenBank mRNA | AF339912.1 | - | 2 | 364 | ||
AF358910.1 | - | 2 | 258 | |||
AK310582.1 | - | 2 | 363 | |||
AK315273.1 | - | 2 | 389 | |||
AY358853.1 | - | 2 | 349 | |||
BC025726.1 | - | 2 | 345 | 5201326 | ||
EU978944.1 | - | 2 | 305 |