Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: EFHC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025184.3 | NP_079460.2 | 5 | 782 | ||
XM_006724562.2 | XP_006724625.1 | 4 | 495 | |||
GenBank mRNA | AK026496.1 | - | 5 | 635 | ||
AK291187.1 | - | 5 | 923 | |||
AK292967.1 | - | 5 | 766 | |||
AK297714.1 | - | 5 | 505 | |||
BC031039.1 | - | 5 | 777 | 5272398 | ||
CR749363.1 | - | 1 | 51 |