Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IER3IP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016097.4 | NP_057181.1 | 1 | 177 | ||
GenBank mRNA | AF125100.1 | - | 1 | 160 | ||
AF164798.1 | - | 1 | 146 | |||
AF371963.1 | - | 1 | 150 | |||
AK027108.1 | - | 1 | 167 | |||
AL136667.1 | - | 1 | 157 | |||
BC010888.1 | - | 1 | 143 | 4285974 | ||
BC017391.1 | - | 1 | 110 | 4133818 | ||
BM829309.1 | - | 1 | 176 | |||
CR533488.1 | - | 1 | 80 |