Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: GPAM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244949.1 | NP_001231878.1 | 3 | 290 | ||
NM_020918.5 | NP_065969.3 | 3 | 286 | |||
XM_005269998.1 | XP_005270055.1 | 3 | 431 | |||
GenBank mRNA | AK172799.1 | - | 1 | 119 | ||
AL832464.1 | - | 3 | 242 | |||
AL833061.2 | - | 3 | 272 | |||
AL833093.2 | - | 3 | 275 | |||
BC030783.1 | - | 3 | 254 | 4821342 | ||
BX647451.1 | - | 3 | 254 | |||
DA313962.1 | - | 3 | 274 | |||
DA802170.1 | - | 3 | 290 | |||
DB197687.1 | - | 3 | 430 |