Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHEB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005614.3 | NP_005605.1 | 3 | 556 | ||
XM_011516457.1 | XP_011514759.1 | 4 | 610 | |||
GenBank mRNA | AF148645.1 | - | 3 | 298 | ||
AF493921.1 | - | 3 | 143 | |||
AK125446.1 | - | 3 | 554 | |||
AK289370.1 | - | 3 | 285 | |||
BC016155.1 | - | 3 | 556 | 3528583 | ||
BC066307.1 | - | 1 | 314 | 5274556 | ||
BC107705.1 | - | 1 | 505 | 6615350 | ||
BG702213.1 | - | 3 | 561 | |||
BT006958.1 | - | 3 | 143 | |||
D78132.1 | - | 3 | 160 | |||
Z29677.1 | - | 3 | 166 |