Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AFF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025108.1 | NP_001020279.1 | 19 | 3227 | ||
NM_002285.2 | NP_002276.2 | 19 | 3244 | |||
XM_005263943.3 | XP_005264000.2 | 18 | 3121 | |||
XM_005263945.3 | XP_005264002.1 | 10 | 2478 | |||
XM_011511169.2 | XP_011509471.1 | 19 | 3224 | |||
XM_011511170.2 | XP_011509472.1 | 20 | 3363 | |||
XM_011511171.2 | XP_011509473.1 | 19 | 3227 | |||
XM_011511172.2 | XP_011509474.1 | 18 | 3179 | |||
XM_011511173.2 | XP_011509475.1 | 19 | 3240 | |||
XM_011511174.2 | XP_011509476.1 | 19 | 3184 | |||
XM_011511175.2 | XP_011509477.1 | 21 | 3474 | |||
XM_011511176.2 | XP_011509478.1 | 18 | 3165 | |||
XM_011511177.2 | XP_011509479.1 | 19 | 3255 | |||
XM_011511178.2 | XP_011509480.1 | 9 | 1986 | |||
XM_011511179.2 | XP_011509481.1 | 9 | 1983 | |||
XM_017004085.1 | XP_016859574.1 | 10 | 2475 | |||
XM_017004086.1 | XP_016859575.1 | 10 | 2956 | |||
XM_017004087.1 | XP_016859576.1 | 10 | 2953 | |||
GenBank mRNA | AB209634.1 | - | 6 | 946 | ||
BC136579.1 | - | 17 | 3027 | 9020569 | ||
BC144266.1 | - | 18 | 3102 | 9052786 | ||
CN409428.1 | - | 1 | 20 | |||
U34360.1 | - | 17 | 3062 |