Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PPP1R16B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172735.2 | NP_001166206.1 | 8 | 1022 | ||
NM_015568.3 | NP_056383.1 | 9 | 1148 | |||
XM_011528768.2 | XP_011527070.1 | 8 | 1711 | |||
XM_011528769.2 | XP_011527071.1 | 7 | 729 | |||
XM_017027785.1 | XP_016883274.1 | 8 | 1043 | |||
GenBank mRNA | AB020630.2 | - | 9 | 1136 | ||
AB177855.1 | - | 7 | 649 | |||
AF362910.1 | - | 8 | 999 | |||
BC131801.1 | - | 9 | 1105 | 40081818 | ||
BC152467.1 | - | 9 | 1136 | 8862603 |