Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DLG2 SIRNA Assays ›
Gene Symbol:
DLG2
Gene Name:
discs large MAGUK scaffold protein 2
Gene Aliases:
PPP1R58, PSD-93, PSD93, chapsyn-110
Chromosome Location:
Chr.11: 83455009 - 85628534 on Build GRCh38
UniGene ID:
Hs.367656
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
44107
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DLG2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001142699.1
NP_001136171.1
28
6087
NM_001142700.1
NP_001136172.1
20
5122
NM_001142702.1
NP_001136174.1
11
4164
NM_001206769.1
NP_001193698.1
22
5771
NM_001300983.1
NP_001287912.1
23
5709
NM_001364.3
NP_001355.2
23
5763
XM_005273810.4
XP_005273867.1
22
5406
XM_005273811.4
XP_005273868.1
22
5352
XM_011544778.2
XP_011543080.1
29
7378
XM_011544780.2
XP_011543082.1
29
7324
XM_011544782.2
XP_011543084.1
24
5849
XM_011544788.2
XP_011543090.1
21
5196
XM_017017254.1
XP_016872743.1
30
7420
XM_017017255.1
XP_016872744.1
29
7381
XM_017017256.1
XP_016872745.1
29
6910
XM_017017257.1
XP_016872746.1
28
7842
XM_017017258.1
XP_016872747.1
27
7018
XM_017017259.1
XP_016872748.1
27
6739
XM_017017260.1
XP_016872749.1
26
7671
XM_017017261.1
XP_016872750.1
28
7168
XM_017017262.1
XP_016872751.1
23
5873
XM_017017263.1
XP_016872752.1
25
5891
XM_017017264.1
XP_016872753.1
24
6068
XM_017017265.1
XP_016872754.1
24
6065
XM_017017266.1
XP_016872755.1
22
5775
XM_017017267.1
XP_016872756.1
24
5973
XM_017017268.1
XP_016872757.1
24
5794
XM_017017269.1
XP_016872758.1
24
6495
XM_017017270.1
XP_016872759.1
24
5795
XM_017017271.1
XP_016872760.1
23
5972
XM_017017272.1
XP_016872761.1
23
5969
XM_017017273.1
XP_016872762.1
22
5724
XM_017017274.1
XP_016872763.1
24
5919
XM_017017275.1
XP_016872764.1
21
5674
XM_017017276.1
XP_016872765.1
23
6397
XM_017017277.1
XP_016872766.1
23
5717
XM_017017278.1
XP_016872767.1
21
5621
XM_017017279.1
XP_016872768.1
23
5639
XM_017017280.1
XP_016872769.1
23
6316
XM_017017281.1
XP_016872770.1
22
5816
XM_017017283.1
XP_016872772.1
22
5621
XM_017017284.1
XP_016872773.1
22
5293
XM_017017285.1
XP_016872774.1
22
6243
XM_017017286.1
XP_016872775.1
20
5157
XM_017017287.1
XP_016872776.1
13
4177
XM_017017288.1
XP_016872777.1
12
4016
XM_017017289.1
XP_016872778.1
12
5591
XM_017017290.1
XP_016872779.1
12
4236
XM_017017291.1
XP_016872780.1
11
3983
XM_017017292.1
XP_016872781.1
11
5495
GenBank mRNA
AK094585.1
-
1
185
AK125151.1
-
1
1193
AK125152.1
-
1
716
BC104969.1
-
1
284
BC110844.1
-
1
539
BX537627.1
-
1
74
BX648386.1
-
1
1590
CR933674.1
-
11
4133
Back To Top
More Information
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
chemical synaptic transmission
nervous system development
negative regulation of phosphatase activity
sensory perception of pain
receptor clustering
establishment or maintenance of epithelial cell apical/basal polarity
GMP metabolic process
GDP metabolic process
receptor localization to synapse
guanylate kinase activity
protein binding
kinase binding
ionotropic glutamate receptor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0