Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WIPI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033518.1 | NP_001028690.1 | 4 | 447 | ||
NM_001033519.1 | NP_001028691.1 | 3 | 393 | |||
NM_001033520.1 | NP_001028692.1 | 1 | 286 | |||
NM_001278299.1 | NP_001265228.1 | 4 | 621 | |||
NM_015610.3 | NP_056425.1 | 4 | 447 | |||
NM_016003.3 | NP_057087.2 | 3 | 393 | |||
XM_006715685.1 | XP_006715748.1 | 2 | 279 | |||
XM_017011952.1 | XP_016867441.1 | 2 | 279 | |||
GenBank mRNA | AF151808.1 | - | 1 | 285 | ||
AK023041.1 | - | 4 | 407 | |||
AK024279.1 | - | 3 | 372 | |||
AK315468.1 | - | 4 | 567 | |||
AL080155.1 | - | 4 | 448 | |||
AY691425.1 | - | 3 | 177 | |||
AY691426.1 | - | 1 | 217 | |||
BC004116.2 | - | 4 | 390 | 3842487 | ||
BC007596.1 | - | 3 | 360 | 3346710 | ||
BC021012.1 | - | 3 | 312 | |||
BC021068.1 | - | 3 | 312 | |||
BC021200.2 | - | 4 | 388 | 3957981 | ||
CR533530.1 | - | 4 | 231 |