Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317812.1 | NP_001304741.1 | 1, 2 | 426 | ||
NM_016612.3 | NP_057696.2 | 1, 2 | 426 | |||
GenBank mRNA | AF223466.1 | - | 1, 2 | 262 | ||
AK074708.1 | - | 1, 2 | 265 | |||
AK075313.1 | - | 1, 2 | 191 | |||
AK223194.1 | - | 1, 2 | 267 | |||
AL833186.1 | - | 1, 2 | 168 | |||
AY032628.1 | - | 1, 2 | 262 | |||
BC015013.2 | - | 1, 2 | 231 | |||
BC132799.1 | - | 1, 2 | 290 | 40146821 | ||
BC132801.1 | - | 1, 2 | 290 | 40146823 | ||
BI460337.1 | - | 1, 2 | 380 | |||
BM926580.1 | - | 1, 2 | 269 | |||
DB057758.1 | - | 1, 2 | 427 |