Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR162 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014449.1 | NP_055264.1 | 3, 4 | 582 | ||
NM_019858.1 | NP_062832.1 | 3, 4 | 1584 | |||
GenBank mRNA | AB209836.1 | - | 2, 3 | 991 | ||
BC002353.2 | - | 3, 4 | 676 | 2821031 | ||
BC007918.2 | - | 2, 3 | 215 | |||
BC080585.1 | - | 3, 4 | 563 | 3510624 | ||
U47925.1 | - | 3, 4 | 582 | |||
U47928.1 | - | 3, 4 | 1584 |