Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PIAS2 SIRNA Assays ›
Gene Symbol:
PIAS2
Gene Name:
protein inhibitor of activated STAT 2
Gene Aliases:
ARIP3, DIP, MIZ, MIZ1, PIASX, PIASX-ALPHA, PIASX-BETA, SIZ2, ZMIZ4
Chromosome Location:
Chr.18: 46803224 - 46920167 on Build GRCh38
UniGene ID:
Hs.57769
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
44313
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PIAS2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001324046.1
NP_001310975.1
4
604
NM_001324047.1
NP_001310976.1
3
515
NM_001324048.1
NP_001310977.1
4
604
NM_001324049.1
NP_001310978.1
5
727
NM_001324051.1
NP_001310980.1
3
515
NM_001324052.1
NP_001310981.1
5
705
NM_001324053.1
NP_001310982.1
5
864
NM_001324054.1
NP_001310983.1
3
495
NM_001324055.1
NP_001310984.1
4
623
NM_001324057.1
NP_001310986.1
5
705
NM_001324058.1
NP_001310987.1
2
406
NM_001324059.1
NP_001310988.1
4
604
NM_001324060.1
NP_001310989.1
2
304
NM_004671.4
NP_004662.2
2
406
NM_173206.3
NP_775298.1
2
406
NR_136684.1
2
406
XM_005258377.3
XP_005258434.1
2
304
XM_005258379.3
XP_005258436.1
2
421
XM_006722571.3
XP_006722634.1
3
389
XM_006722572.3
XP_006722635.1
3
389
XM_006722573.2
XP_006722636.1
3
389
XM_011526258.2
XP_011524560.1
2
469
XM_011526259.2
XP_011524561.1
3
320
XM_017026068.1
XP_016881557.1
5
705
XM_017026069.1
XP_016881558.1
3
438
XM_017026070.1
XP_016881559.1
5
705
XM_017026071.1
XP_016881560.1
3
320
XM_017026072.1
XP_016881561.1
2
259
GenBank mRNA
AF077953.1
-
2
219
AF077954.1
-
2
219
AF361054.1
-
2
297
AK308143.1
-
2
328
BC015190.1
-
2
377
4080297
BC111060.1
-
2
348
5745101
BG718291.1
-
2
413
BX646432.1
-
2
380
DB024780.1
-
2
304
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
protein sumoylation
androgen receptor signaling pathway
regulation of osteoblast differentiation
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
negative regulation of androgen receptor signaling pathway
DNA binding
transcription coactivator activity
protein binding
transcription factor binding
zinc ion binding
ligase activity
SUMO transferase activity
ubiquitin protein ligase binding
androgen receptor binding
SUMO ligase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-574f4745d9-ftf7j:80/10.239.160.32:80.
git-commit: d20c60ba4e48417b3d41b4fa5195ecf3ea66aacf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.12.0-2024.07.30-1.0