Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other KCNIP4 SIRNA Assays ›
Gene Symbol:
KCNIP4
Gene Name:
potassium voltage-gated channel interacting protein 4
Gene Aliases:
CALP, KCHIP4
Chromosome Location:
Chr.4: 20728520 - 21948801 on Build GRCh38
UniGene ID:
Hs.655705
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
44386
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: KCNIP4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001035003.1
NP_001030175.1
3, 4
545
NM_001035004.1
NP_001030176.1
3, 4
466
NM_025221.5
NP_079497.2
4, 5
482
NM_147181.3
NP_671710.1
3, 4
380
NM_147182.3
NP_671711.1
4, 5
492
NM_147183.3
NP_671712.1
3, 4
299
XM_005248190.4
XP_005248247.1
3, 4
395
XM_011513882.2
XP_011512184.1
4, 5
526
XM_011513883.2
XP_011512185.1
3, 4
395
XM_011513884.2
XP_011512186.1
3, 4
419
XM_011513885.2
XP_011512187.1
3, 4
5715
XM_011513886.2
XP_011512188.1
3, 4
430
XM_011513887.2
XP_011512189.1
3, 4
1195
XM_011513889.2
XP_011512191.1
3, 4
385
XM_017008653.1
XP_016864142.1
3, 4
568
GenBank mRNA
AF302044.1
-
2, 3
260
AF305072.1
-
3, 4
362
AF367023.1
-
3, 4
412
AF367024.1
-
2, 3
310
AF453246.1
-
2, 3
176
AK289661.1
-
3, 4
374
AK289922.1
-
4, 5
452
AK290998.1
-
3, 4
382
AK294188.1
-
3, 4
395
AK314058.1
-
546
AY029176.1
-
3, 4
372
AY118170.1
-
3, 4
299
BC032520.1
-
3, 4
420
5494835
BI596848.1
-
487
BX648990.1
-
3, 4
546
DQ148487.1
-
3, 4
362
DQ148488.1
-
2, 3
260
DQ148489.1
-
3, 4
299
DQ148490.1
-
3, 4
545
DQ148491.1
-
3, 4
466
DQ148492.1
-
3, 4
311
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
cardiac conduction
potassium ion transmembrane transport
protein localization to plasma membrane
regulation of potassium ion transmembrane transport
voltage-gated ion channel activity
potassium channel activity
calcium ion binding
potassium channel regulator activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-4vtwv:80/10.239.167.181:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline