Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001280796.1 | NP_001267725.1 | 8, 9 | 1162 | ||
NM_001280797.1 | NP_001267726.1 | 7, 8 | 1032 | |||
NM_001280798.1 | NP_001267727.1 | 5, 6 | 810 | |||
NM_013267.3 | NP_037399.2 | 8, 9 | 1162 | |||
XM_005268797.1 | XP_005268854.1 | 8, 9 | 789 | |||
XM_017019180.1 | XP_016874669.1 | 7, 8 | 659 | |||
GenBank mRNA | AF110330.1 | - | 8, 9 | 917 | ||
AF223944.1 | - | 8, 9 | 1089 | |||
AK000467.1 | - | 8, 9 | 909 | |||
BC025661.1 | - | 7, 8 | 747 | |||
DA060302.1 | - | 5, 6 | 394 | |||
HM775422.1 | - | 7, 8 | 781 | |||
HM775423.1 | - | 6, 7 | 651 | |||
HM775424.1 | - | 7, 8 | 781 |