Search Thermo Fisher Scientific
- ์ฃผ๋ฌธ ํํฉ
- ๋น ๋ฅธ์ฃผ๋ฌธ
-
ํ์์ด ์๋์ ๊ฐ์? ๊ณ์ ์์ฑํ๊ธฐ
Search Thermo Fisher Scientific
Gene Symbol: WSB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015626.8 | NP_056441.6 | 8 | 1350 | ||
NM_134265.2 | NP_599027.1 | 6 | 912 | |||
XM_005257963.3 | XP_005258020.1 | 7 | 1147 | |||
XM_017024433.1 | XP_016879922.1 | 7 | 1236 | |||
XM_017024434.1 | XP_016879923.1 | 9 | 1438 | |||
XM_017024435.1 | XP_016879924.1 | 9 | 2805 | |||
XM_017024436.1 | XP_016879925.1 | 7 | 2537 | |||
GenBank mRNA | AF069313.2 | - | 8 | 1299 | ||
AF072880.1 | - | 8 | 1125 | |||
AF106683.1 | - | 8 | 1167 | |||
AF106684.1 | - | 3 | 899 | |||
AF112205.1 | - | 8 | 1289 | |||
AF240696.1 | - | 6 | 861 | |||
AK074917.1 | - | 8 | 1316 | |||
AK222576.1 | - | 8 | 1316 | |||
AL110243.1 | - | 8 | 1350 | |||
AL110269.1 | - | 8 | 2677 | |||
BC005226.1 | - | 3 | 352 | |||
BC021110.1 | - | 8 | 1297 | 4865448 | ||
BC048007.1 | - | 8 | 2672 |