Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317812.1 | NP_001304741.1 | 4 | 1716 | ||
NM_001317813.1 | NP_001304742.1 | 5 | 1363 | |||
NM_001317814.1 | NP_001304743.1 | 5 | 1310 | |||
NM_016612.3 | NP_057696.2 | 4 | 1239 | |||
XM_006716352.3 | XP_006716415.1 | 4 | 843 | |||
XM_011544550.2 | XP_011542852.1 | 4 | 843 | |||
XM_011544553.2 | XP_011542855.1 | 5 | 2075 | |||
XM_011544554.2 | XP_011542856.1 | 4 | 2997 | |||
XM_017013532.1 | XP_016869021.1 | 4 | 1914 | |||
GenBank mRNA | AF155660.1 | - | 2 | 1057 | ||
AF223466.1 | - | 4 | 1074 | |||
AK074708.1 | - | 4 | 1555 | |||
AK075313.1 | - | 4 | 1480 | |||
AK093931.1 | - | 1 | 2341 | |||
AK127666.1 | - | 4 | 3308 | |||
AK223194.1 | - | 4 | 1556 | |||
AY032628.1 | - | 3 | 3062 | |||
BC132799.1 | - | 4 | 1103 | 40146821 | ||
BC132801.1 | - | 4 | 1103 | 40146823 |