Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TRERF1 SIRNA Assays ›
Gene Symbol:
TRERF1
Gene Name:
transcriptional regulating factor 1
Gene Aliases:
BCAR2, HSA277276, RAPA, TREP132, TReP-132, dJ139D8.5
Chromosome Location:
Chr.6: 42224933 - 42452216 on Build GRCh38
UniGene ID:
Hs.485392
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
44488
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TRERF1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001297573.1
NP_001284502.1
13
3255
NM_033502.3
NP_277037.1
13
3195
XM_011514741.2
XP_011513043.1
13
3766
XM_011514742.2
XP_011513044.1
15
3942
XM_011514743.2
XP_011513045.1
14
3865
XM_011514744.1
XP_011513046.1
13
3374
XM_011514745.1
XP_011513047.1
13
3268
XM_017011034.1
XP_016866523.1
13
3816
XM_017011035.1
XP_016866524.1
13
3484
XM_017011036.1
XP_016866525.1
13
3303
XM_017011037.1
XP_016866526.1
13
3334
XM_017011038.1
XP_016866527.1
15
3956
XM_017011039.1
XP_016866528.1
13
3561
XM_017011040.1
XP_016866529.1
13
3253
XM_017011041.1
XP_016866530.1
13
3375
XM_017011042.1
XP_016866531.1
12
4478
XM_017011043.1
XP_016866532.1
12
5488
XM_017011044.1
XP_016866533.1
13
3765
XM_017011045.1
XP_016866534.1
13
3705
XM_017011046.1
XP_016866535.1
13
3673
XM_017011047.1
XP_016866536.1
13
3627
XM_017011048.1
XP_016866537.1
13
3673
XM_017011049.1
XP_016866538.1
13
3613
XM_017011050.1
XP_016866539.1
13
3766
XM_017011051.1
XP_016866540.1
12
3364
XM_017011052.1
XP_016866541.1
12
3364
XM_017011053.1
XP_016866542.1
14
3540
XM_017011054.1
XP_016866543.1
12
3364
XM_017011055.1
XP_016866544.1
12
2072
GenBank mRNA
AF297872.1
-
13
3189
AJ277275.1
-
12
2730
AJ277276.1
-
12
2730
AK127493.1
-
9
2669
AM404182.1
-
13
3255
AM404183.1
-
13
3195
AM404259.1
-
13
3195
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001108199, XM_006244452, XM_008766863, XM_006244453.This siRNA also targets the following transcripts in Mouse: NM_172622, NM_001097623, AK035176, AY081774, AK046822, and others.
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
steroid biosynthetic process
cholesterol catabolic process
multicellular organism development
homeostatic process
positive regulation of transcription, DNA-templated
regulation of hormone biosynthetic process
RNA polymerase II transcription cofactor activity
transcription factor activity, sequence-specific DNA binding
transcription factor binding
DNA binding, bending
ligand-dependent nuclear receptor transcription coactivator activity
transcription regulatory region DNA binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-qvdz4:80/10.239.165.207:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0