Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARID1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017519.2 | NP_059989.2 | 2 | 1580 | ||
NM_020732.3 | NP_065783.3 | 2 | 1580 | |||
XM_005267069.4 | XP_005267126.2 | 2 | 1655 | |||
XM_011535984.2 | XP_011534286.2 | 2 | 1656 | |||
XM_017011103.1 | XP_016866592.1 | 2 | 1656 | |||
XM_017011104.1 | XP_016866593.1 | 2 | 1655 | |||
XM_017011105.1 | XP_016866594.1 | 2 | 1656 | |||
XM_017011106.1 | XP_016866595.1 | 2 | 1655 | |||
XM_017011107.1 | XP_016866596.1 | 2 | 1656 | |||
XM_017011108.1 | XP_016866597.1 | 2 | 1663 | |||
GenBank mRNA | AF253515.1 | - | 2 | 774 | ||
AF468300.1 | - | 2 | 110 | |||
AF521671.1 | - | 2 | 1367 |