Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001022.3 | NP_001013.1 | 2, 3 | 452 | ||
NM_001321483.1 | NP_001308412.1 | 2, 3 | 129 | |||
NM_001321484.1 | NP_001308413.1 | 2, 3 | 259 | |||
NM_001321485.1 | NP_001308414.1 | 2, 3 | 452 | |||
XM_017027113.1 | XP_016882602.1 | 2, 3 | 447 | |||
GenBank mRNA | AK311786.1 | - | 2, 3 | 115 | ||
BC000023.2 | - | 2, 3 | 149 | 3504083 | ||
BC007615.2 | - | 1, 2 | 80 | 3354280 | ||
BC017386.1 | - | 2, 3 | 118 | |||
BC018616.1 | - | 2, 3 | 452 | 4867036 | ||
BG398397.1 | - | 2, 3 | 259 | |||
BM468237.1 | - | 2, 3 | 223 | |||
BP270469.1 | - | 2, 3 | 115 | |||
BU603942.1 | - | 2, 3 | 118 | |||
CB124389.1 | - | 2, 3 | 158 | |||
CB125125.1 | - | 2, 3 | 129 | |||
M81757.1 | - | 2, 3 | 102 |