Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HCRT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001524.1 | NP_001515.1 | 2 | 556 | ||
GenBank mRNA | AF041240.1 | - | 2 | 556 |