Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEACAM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277163.2 | NP_001264092.1 | 2, 3 | 507 | ||
NM_001815.4 | NP_001806.2 | 2, 3 | 507 | |||
GenBank mRNA | BC106728.1 | - | 2, 3 | 445 | 40015072 | |
D90277.1 | - | 2, 3 | 493 | |||
E03348.1 | - | 2, 3 | 493 | |||
L00692.1 | - | 2, 3 | 473 | |||
L00693.1 | - | 2, 3 | 473 |