Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNRPB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003091.3 | NP_003082.1 | 2, 3 | 321 | ||
NM_198216.1 | NP_937859.1 | 2, 3 | 321 | |||
GenBank mRNA | AK301206.1 | - | 2, 3 | 255 | ||
AK312620.1 | - | 2, 3 | 256 | |||
AY742712.1 | - | 1, 2 | 158 | |||
BC080516.1 | - | 2, 3 | 239 | 2822685 | ||
BM559678.1 | - | 321 | ||||
CR456969.1 | - | 1, 2 | 158 | |||
J04564.1 | - | 2, 3 | 284 | |||
M34081.1 | - | 1, 2 | 137 | |||
M34082.1 | - | 1, 2 | 137 | |||
X15893.1 | - | 1, 2 | 158 | |||
X17567.1 | - | 2, 3 | 212 | |||
X17568.1 | - | 2, 3 | 197 |