Gene Symbol: SOAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252511.1 | NP_001239440.1 | 2 | 332 | ||
NM_003101.5 | NP_003092.4 | 2 | 332 | |||
XM_011509911.1 | XP_011508213.1 | 2 | 520 | |||
XM_011509912.1 | XP_011508214.1 | 2 | 327 | |||
GenBank mRNA | AK026611.1 | - | 2 | 244 | ||
AK290659.1 | - | 2 | 249 | |||
AL833625.1 | - | 2 | 237 | |||
BC028940.1 | - | 2 | 164 | 4991365 | ||
DC344271.1 | - | 2 | 332 | |||
L21934.2 | - | 1497 |