Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other GAPDH SIRNA Assays ›
Gene Symbol:
GAPDH
Gene Name:
glyceraldehyde-3-phosphate dehydrogenase
Gene Aliases:
G3PD, GAPD, HEL-S-162eP
Chromosome Location:
Chr.12: 6534405 - 6538375 on Build GRCh38
UniGene ID:
Hs.544577
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
44902
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: GAPDH
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001256799.2
NP_001243728.1
3
413
NM_001289745.1
NP_001276674.1
4
471
NM_001289746.1
NP_001276675.1
3
365
NM_002046.5
NP_002037.2
4
379
GenBank mRNA
AB062273.1
-
4
243
AF261085.1
-
4
294
AJ844644.1
-
2
109
AK026525.1
-
4
316
AK299448.1
-
4
264
AK308198.1
-
4
267
AY007133.1
-
4
243
AY633612.1
-
3
215
BC001601.1
-
4
245
3543589
BC004109.2
-
4
243
3678259
BC009081.1
-
4
252
3869809
BC013310.2
-
4
251
3538383
BC020308.1
-
3
229
BC023632.2
-
3
215
4842418
BC025925.1
-
4
248
2989348
BC026907.1
-
3
229
4454083
BC029340.1
-
4
237
BC029618.1
-
4
245
3876535
BC083511.1
-
4
243
3536969
BG724119.1
-
3
396
BI463134.1
-
3
395
BM763361.1
-
4
356
BT006893.1
-
3
191
BU155402.1
-
4
416
CR407671.1
-
3
191
DQ403057.1
-
2
116
EU668321.1
-
4
293
HQ727992.1
-
2
81
HY000136.1
-
3
414
HY004110.1
-
3
366
HY022295.1
-
3
342
J02642.1
-
3
192
M17851.1
-
3
224
M33197.1
-
4
251
X01677.1
-
3
251
X53778.1
-
4
247
Back To Top
More Information
Panther Classification:
Molecular Function -
dehydrogenase
Gene Ontology Categories:
Function(s)
Process(es)
microtubule cytoskeleton organization
gluconeogenesis
regulation of macroautophagy
negative regulation of translation
peptidyl-cysteine S-trans-nitrosylation
protein stabilization
neuron apoptotic process
canonical glycolysis
cellular response to interferon-gamma
glyceraldehyde-3-phosphate dehydrogenase (NAD+) (phosphorylating) activity
protein binding
microtubule binding
peptidyl-cysteine S-nitrosylase activity
identical protein binding
NADP binding
NAD binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-8gqds:80/10.239.164.183:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0