Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ICAM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320605.1 | NP_001307534.1 | 2 | 411 | ||
NM_001320606.1 | NP_001307535.1 | 2 | 708 | |||
NM_001320608.1 | NP_001307537.1 | 2 | 411 | |||
NM_002162.4 | NP_002153.2 | 2 | 411 | |||
GenBank mRNA | AF340060.1 | - | 2 | 164 | ||
AF340061.1 | - | 2 | 164 | |||
AK226023.1 | - | 2 | 245 | |||
AK301071.1 | - | 2 | 116 | |||
AK309643.1 | - | 2 | 493 | |||
AK312826.1 | - | 2 | 246 | |||
AX746321.1 | - | 2 | 206 | |||
BC040123.1 | - | 2 | 191 | |||
BC046121.1 | - | 2 | 238 | |||
BC058903.1 | - | 2 | 197 | 5207143 | ||
CR982084.1 | - | 2 | 411 | |||
DC425414.1 | - | 2 | 400 | |||
S50015.1 | - | 2 | 192 | |||
X69711.1 | - | 2 | 206 | |||
X69819.1 | - | 2 | 199 |