Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286356.1 | NP_001273285.1 | 2 | 450 | ||
NM_001286358.1 | NP_001273287.1 | 2 | 399 | |||
NM_001286359.1 | NP_001273288.1 | 2 | 453 | |||
NM_002762.3 | NP_002753.2 | 2 | 406 | |||
NR_104428.1 | 2 | 406 | ||||
GenBank mRNA | AK130957.1 | - | 2 | 406 | ||
AK131573.1 | - | 2 | 450 | |||
AK292447.1 | - | 2 | 406 | |||
AK311798.1 | - | 2 | 406 | |||
BC005303.1 | - | 2 | 406 | 3935273 | ||
BC042671.1 | - | 2 | 375 | 5166075 | ||
BC066338.1 | - | 2 | 406 | 4821351 | ||
BP368978.1 | - | 2 | 406 | |||
DB066857.1 | - | 2 | 403 | |||
DB066922.1 | - | 2 | 454 | |||
X07862.1 | - | 2 | 377 |