Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004123.2 | NP_004114.1 | 2 | 170 | ||
GenBank mRNA | BC069100.1 | - | 2 | 170 | 7216914 | |
BC069663.1 | - | 2 | 170 | 7262542 | ||
BC069686.1 | - | 2 | 170 | 7262554 | ||
BC069746.1 | - | 2 | 170 | 7262566 | ||
BC096146.1 | - | 2 | 169 | 40002916 | ||
BC096147.1 | - | 2 | 169 | 40002922 | ||
BC096148.3 | - | 2 | 169 | 40002923 | ||
BC096149.3 | - | 2 | 169 | 40002925 | ||
E02006.1 | - | 2 | 172 | |||
M18185.1 | - | 2 | 170 |