Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FEZ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005103.4 | NP_005094.1 | 2 | 378 | ||
NM_022549.3 | NP_072043.1 | 2 | 378 | |||
XM_005271734.2 | XP_005271791.1 | 2 | 423 | |||
XM_005271735.2 | XP_005271792.1 | 2 | 475 | |||
GenBank mRNA | AK296554.1 | - | 2 | 285 | ||
BC009545.1 | - | 2 | 295 | 3897353 | ||
CN310685.1 | - | 2 | 320 | |||
CR456817.1 | - | 1 | 143 | |||
CR541954.1 | - | 1 | 143 | |||
DA697171.1 | - | 2 | 385 | |||
U60060.1 | - | 2 | 242 | |||
U60062.1 | - | 1 | 172 | |||
U69139.1 | - | 1 | 168 |