Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TCF4 SIRNA Assays ›
Gene Symbol:
TCF4
Gene Name:
transcription factor 4
Gene Aliases:
E2-2, FECD3, ITF-2, ITF2, PTHS, SEF-2, SEF2, SEF2-1, SEF2-1A, SEF2-1B, SEF2-1D, TCF-4, bHLHb19
Chromosome Location:
Chr.18: 55222331 - 55635993 on Build GRCh38
UniGene ID:
Hs.605153
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
4511
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TCF4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001083962.1
NP_001077431.1
4, 5
826
NM_001243226.2
NP_001230155.2
5, 6
616
NM_001243227.1
NP_001230156.1
3, 4
660
NM_001243228.1
NP_001230157.1
4, 5
752
NM_001243230.1
NP_001230159.1
3, 4
302
NM_001243231.1
NP_001230160.1
2, 3
260
NM_001306207.1
NP_001293136.1
3, 4
451
NM_003199.2
NP_003190.1
4, 5
826
XM_005266739.3
XP_005266796.2
3, 4
466
XM_005266741.3
XP_005266798.1
4, 5
429
XM_005266743.4
XP_005266800.1
3, 4
287
XM_005266744.3
XP_005266801.1
3, 4
407
XM_005266745.3
XP_005266802.1
3, 4
272
XM_005266747.3
XP_005266804.1
2, 3
263
XM_006722536.2
XP_006722599.1
4, 5
749
XM_006722537.2
XP_006722600.1
4, 5
382
XM_006722538.2
XP_006722601.1
3, 4
407
XM_011526158.2
XP_011524460.1
3, 4
1380
XM_017025934.1
XP_016881423.1
3, 4
466
XM_017025935.1
XP_016881424.1
3, 4
466
XM_017025936.1
XP_016881425.1
3, 4
466
XM_017025937.1
XP_016881426.1
3, 4
223
XM_017025938.1
XP_016881427.1
4, 5
382
XM_017025939.1
XP_016881428.1
4, 5
429
XM_017025940.1
XP_016881429.1
4, 5
749
XM_017025941.1
XP_016881430.1
4, 5
382
XM_017025942.1
XP_016881431.1
4, 5
429
XM_017025943.1
XP_016881432.1
4, 5
429
XM_017025944.1
XP_016881433.1
3, 4
687
XM_017025945.1
XP_016881434.1
3, 4
687
XM_017025946.1
XP_016881435.1
3, 4
701
XM_017025947.1
XP_016881436.1
3, 4
287
XM_017025948.1
XP_016881437.1
3, 4
687
XM_017025949.1
XP_016881438.1
3, 4
407
XM_017025950.1
XP_016881439.1
3, 4
687
XM_017025951.1
XP_016881440.1
2, 3
263
GenBank mRNA
AK096862.1
-
2, 3
260
AK122765.1
-
4, 5
826
AK299169.1
-
3, 4
660
AK301144.1
-
3, 4
437
AK315074.1
-
4, 5
740
BC031056.1
-
5, 6
577
BC125084.1
-
3, 4
240
40117795
BC125085.1
-
3, 4
240
40117798
BP293388.1
-
3, 4
289
FR748205.1
-
4, 5
288
FR748207.1
-
2, 3
144
FR748210.1
-
3, 4
214
FR748211.1
-
3, 4
214
FR748212.1
-
3, 4
214
FR748213.1
-
3, 4
214
FR748214.1
-
2, 3
142
FR748215.1
-
2, 3
142
FR748216.1
-
3, 4
208
FR748217.1
-
2, 3
88
M74719.1
-
4, 5
413
X52079.1
-
1, 2
79
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_053369, AF149284.This siRNA also targets the following transcripts in Mouse: NM_013685, NM_001083967, AK133885, AK028748, AK081012, and others.
Panther Classification:
Molecular Function -
basic helix-loop-helix transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
DNA-templated transcription, initiation
transcription initiation from RNA polymerase II promoter
positive regulation of neuron differentiation
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
protein-DNA complex assembly
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcription factor activity, sequence-specific DNA binding, RNA polymerase recruiting
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcription factor activity, TFIIB-class binding
TFIIB-class transcription factor binding
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
protein C-terminus binding
identical protein binding
protein homodimerization activity
bHLH transcription factor binding
protein heterodimerization activity
E-box binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-jmrqp:80/10.239.165.107:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0