Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: MAGEA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005365.4 | NP_005356.1 | 4 | 542 | ||
XM_005262334.3 | XP_005262391.1 | 4 | 564 | |||
XM_005262335.3 | XP_005262392.1 | 3 | 471 | |||
GenBank mRNA | AK292272.1 | - | 4 | 549 | ||
AK313321.1 | - | 4 | 549 | |||
BC002351.2 | - | 4 | 542 | 2820966 | ||
BC098100.1 | - | 4 | 503 |
Gene Symbol: MAGEA9B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080790.1 | NP_001074259.1 | 4 | 549 | ||
XM_005278192.3 | XP_005278249.1 | 4 | 564 | |||
XM_005278193.3 | XP_005278250.1 | 3 | 471 |