Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAGEA12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166386.3 | NP_001159858.1 | 3 | 633 | ||
NM_001166387.3 | NP_001159859.1 | 3 | 541 | |||
NM_005367.6 | NP_005358.2 | 2 | 435 | |||
GenBank mRNA | AK314168.1 | - | 3 | 633 | ||
BC003408.2 | - | 2 | 423 | 3450217 | ||
BC098146.1 | - | 3 | 213 | |||
BQ436888.1 | - | 2 | 435 | |||
BT007108.1 | - | 1 | 279 | |||
CR541775.1 | - | 1 | 279 |