Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NR2F1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005654.5 | NP_005645.1 | 1 | 1983 | ||
GenBank mRNA | BC004154.2 | - | 1 | 384 | 2824138 | |
BC017493.1 | - | 1 | 1983 | 3912075 | ||
HQ709180.1 | - | 2 | 221 | |||
HQ709181.1 | - | 2 | 143 | |||
HQ709182.1 | - | 2 | 173 | |||
HQ709183.1 | - | 2 | 221 | |||
X16155.1 | - | 1 | 281 |