Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CCL7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006273.3 | NP_006264.2 | 1, 2 | 155 | ||
GenBank mRNA | BC092436.1 | - | 1, 2 | 149 | 30418073 | |
BC112258.1 | - | 1, 2 | 145 | 8327726 | ||
BC112260.1 | - | 1, 2 | 145 | 8327728 | ||
CD523198.1 | - | 1, 2 | 158 | |||
X71087.1 | - | 1, 2 | 150 |