Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TLE4 SIRNA Assays ›
Gene Symbol:
TLE4
Gene Name:
transducin like enhancer of split 4
Gene Aliases:
BCE-1, BCE1, E(spI), ESG, ESG4, GRG4, Grg-4
Chromosome Location:
Chr.9: 79571773 - 79726882 on Build GRCh38
UniGene ID:
Hs.444213
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
45167
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TLE4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001282748.1
NP_001269677.1
21
4439
NM_001282749.1
NP_001269678.1
19
4268
NM_001282753.1
NP_001269682.1
18
4136
NM_001282760.1
NP_001269689.1
21
4438
NM_007005.4
NP_008936.2
20
4343
NR_104239.1
21
4476
XM_005252177.1
XP_005252234.1
20
3519
XM_006717264.1
XP_006717327.1
23
3738
XM_006717268.1
XP_006717331.1
22
3642
XM_011518952.1
XP_011517254.1
24
3813
XM_011518953.1
XP_011517255.1
24
3690
XM_011518954.1
XP_011517256.1
23
3768
XM_011518955.1
XP_011517257.1
23
3750
XM_011518956.1
XP_011517258.1
23
3729
XM_011518957.1
XP_011517259.1
23
3726
XM_011518958.1
XP_011517260.1
23
3717
XM_011518959.1
XP_011517261.1
22
3690
XM_011518960.1
XP_011517262.1
22
3687
XM_011518961.1
XP_011517263.1
21
3627
XM_011518962.1
XP_011517264.1
21
3624
XM_011518963.1
XP_011517265.1
21
3615
XM_011518964.1
XP_011517266.1
21
3612
XM_011518965.1
XP_011517267.1
21
3594
XM_011518966.1
XP_011517268.1
21
3591
XM_011518967.1
XP_011517269.1
22
3564
XM_011518968.1
XP_011517270.1
20
3552
XM_011518969.1
XP_011517271.1
21
3510
XM_011518972.2
XP_011517274.1
12
2640
XM_017015067.1
XP_016870556.1
22
3618
XM_017015068.1
XP_016870557.1
21
3645
XM_017015069.1
XP_016870558.1
21
3473
XM_017015070.1
XP_016870559.1
21
3550
XM_017015071.1
XP_016870560.1
21
3555
XM_017015072.1
XP_016870561.1
20
3476
XM_017015073.1
XP_016870562.1
20
3478
XM_017015074.1
XP_016870563.1
21
3489
XM_017015075.1
XP_016870564.1
19
3471
XM_017015076.1
XP_016870565.1
19
3418
XM_017015077.1
XP_016870566.1
19
3456
XM_017015078.1
XP_016870567.1
19
3453
XM_017015079.1
XP_016870568.1
19
3381
XM_017015080.1
XP_016870569.1
19
3444
XM_017015081.1
XP_016870570.1
20
3441
XM_017015082.1
XP_016870571.1
19
3336
XM_017015083.1
XP_016870572.1
19
3363
XM_017015084.1
XP_016870573.1
18
3288
XM_017015085.1
XP_016870574.1
18
3196
XM_017015086.1
XP_016870575.1
17
3234
GenBank mRNA
AF068197.1
-
1
595
BC060038.1
-
1
813
W68064.1
-
1
307
Back To Top
More Information
Panther Classification:
Molecular Function -
transcription cofactor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
biological_process
Wnt signaling pathway
beta-catenin-TCF complex assembly
molecular_function
chromatin binding
transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
transcription corepressor activity
protein binding
repressing transcription factor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-46vz9:80/10.239.163.144:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0