Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPY5R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317091.1 | NP_001304020.1 | 4 | 1653 | ||
NM_001317092.1 | NP_001304021.1 | 5 | 533 | |||
NM_006174.3 | NP_006165.1 | 4 | 405 | |||
XM_005263038.3 | XP_005263095.1 | 4 | 7287 | |||
XM_011532015.2 | XP_011530317.1 | 5 | 7324 | |||
XM_011532017.2 | XP_011530319.1 | 5 | 7415 | |||
XM_017008255.1 | XP_016863744.1 | 5 | 365 | |||
XM_017008256.1 | XP_016863745.1 | 6 | 7452 | |||
GenBank mRNA | BC042416.1 | - | 4 | 313 | 5191617 | |
U56079.1 | - | 1 | 186 | |||
U66275.1 | - | 1 | 148 | |||
U94320.1 | - | 2 | 201 |
Set Membership: |
G-protein coupled receptor |