Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: YPEL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013313.4 | NP_037445.1 | 4, 5 | 665 | ||
NR_130910.1 | 4, 5 | 1049 | ||||
GenBank mRNA | AB098734.1 | - | 4, 5 | 665 | ||
AB098735.1 | - | 4, 5 | 665 | |||
AF060862.1 | - | 3, 4 | 365 | |||
AF305069.1 | - | 3, 4 | 425 | |||
BC034486.1 | - | 4, 5 | 523 | 4826997 | ||
BC074501.1 | - | 4, 5 | 530 | 5171826 | ||
BI559478.1 | - | 4, 5 | 545 | |||
CR456376.2 | - | 3, 4 | 312 | |||
CR933630.1 | - | 3, 4 | 4768 |