Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RIPK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317061.1 | NP_001303990.1 | 4 | 593 | ||
NM_003804.4 | NP_003795.2 | 3 | 429 | |||
XM_005249457.3 | XP_005249514.2 | 3 | 352 | |||
XM_006715237.3 | XP_006715300.2 | 3 | 352 | |||
XM_017011405.1 | XP_016866894.1 | 3 | 350 | |||
GenBank mRNA | AB208926.1 | - | 4 | 502 | ||
AK304701.1 | - | 2 | 156 | |||
AK314176.1 | - | 3 | 429 | |||
BC126254.1 | - | 2 | 223 | 8991970 | ||
BC126256.1 | - | 2 | 223 | 8991972 | ||
U50062.1 | - | 2 | 197 |