Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LY6G6E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003673.3 | 2 | 412 | |||
NR_024541.1 | 2 | 412 | ||||
GenBank mRNA | AJ245419.1 | - | 2 | 159 | ||
AJ315538.1 | - | 2 | 159 | |||
AJ315539.1 | - | 2 | 159 | |||
BC069141.1 | - | 2 | 159 | |||
DA786728.1 | - | 2 | 412 |