Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OGG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002542.5 | NP_002533.1 | 2 | 538 | ||
NM_016819.3 | NP_058212.1 | 2 | 538 | |||
NM_016820.3 | NP_058213.1 | 2 | 538 | |||
NM_016821.2 | NP_058214.1 | 2 | 538 | |||
NM_016826.2 | NP_058434.1 | 2 | 538 | |||
NM_016827.2 | NP_058436.1 | 2 | 538 | |||
NM_016828.2 | NP_058437.1 | 2 | 538 | |||
NM_016829.2 | NP_058438.1 | 2 | 538 | |||
XM_011533760.1 | XP_011532062.1 | 2 | 533 | |||
XM_017006493.1 | XP_016861982.1 | 2 | 533 | |||
XM_017006494.1 | XP_016861983.1 | 2 | 533 | |||
XM_017006495.1 | XP_016861984.1 | 2 | 533 | |||
XM_017006496.1 | XP_016861985.1 | 2 | 533 | |||
XM_017006497.1 | XP_016861986.1 | 2 | 533 | |||
XM_017006498.1 | XP_016861987.1 | 2 | 533 | |||
XM_017006499.1 | XP_016861988.1 | 2 | 533 | |||
GenBank mRNA | AB000410.1 | - | 2 | 463 | ||
AF003595.1 | - | 2 | 251 | |||
AF026691.1 | - | 2 | 402 | |||
AK289858.1 | - | 2 | 491 | |||
BC000657.2 | - | 2 | 492 | 3350168 | ||
BM847736.1 | - | 2 | 539 | |||
U88527.1 | - | 2 | 505 | |||
U88620.1 | - | 2 | 505 | |||
U96710.1 | - | 2 | 252 | |||
Y11731.2 | - | 2 | 503 | |||
Y11838.1 | - | 2 | 502 | |||
Y13277.1 | - | 2 | 499 |