Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF91 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003430.3 | NP_003421.2 | 3, 4 | 467 | ||
XM_011528265.2 | XP_011526567.1 | 3, 4 | 428 | |||
XM_011528266.2 | XP_011526568.1 | 4 | 396 | |||
XM_017027235.1 | XP_016882724.1 | 3, 4 | 421 | |||
XM_017027236.1 | XP_016882725.1 | 2, 3 | 288 | |||
GenBank mRNA | AK291256.1 | - | 3, 4 | 374 | ||
DB068854.1 | - | 3, 4 | 467 | |||
DB219374.1 | - | 3, 4 | 374 | |||
L11672.1 | - | 3, 4 | 355 | |||
M61871.1 | - | 2, 3 | 219 |