Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AHSA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321300.1 | NP_001308229.1 | 5 | 1428 | ||
NM_001321301.1 | NP_001308230.1 | 5 | 2140 | |||
NM_001321302.1 | NP_001308231.1 | 6 | 848 | |||
NM_001321303.1 | NP_001308232.1 | 4 | 2024 | |||
NM_001321304.1 | NP_001308233.1 | 4 | 2024 | |||
NM_001321305.1 | NP_001308234.1 | 4 | 2024 | |||
NM_152392.4 | NP_689605.1 | 4 | 2024 | |||
XM_011532541.2 | XP_011530843.1 | 5 | 982 | |||
XM_011532543.2 | XP_011530845.1 | 6 | 1266 | |||
XM_011532545.2 | XP_011530847.1 | 5 | 982 | |||
XM_017003377.1 | XP_016858866.1 | 5 | 982 | |||
XM_017003378.1 | XP_016858867.1 | 6 | 941 | |||
GenBank mRNA | AF542548.1 | - | 3 | 225 | ||
AK092505.1 | - | 4 | 411 | |||
AK092817.1 | - | 4 | 1428 | |||
AL833813.1 | - | 4 | 1759 | |||
BC063789.1 | - | 4 | 1913 | |||
BC113671.1 | - | 4 | 1021 | 8322723 | ||
BC117278.1 | - | 4 | 1021 | 40125829 |