Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173505.3 | NP_775776.2 | 9 | 914 | ||
XM_011525830.2 | XP_011524132.1 | 8 | 834 | |||
XM_011525831.2 | XP_011524133.1 | 8 | 2854 | |||
XM_011525832.2 | XP_011524134.1 | 8 | 2972 | |||
GenBank mRNA | AK123197.1 | - | 6 | 2234 | ||
AK313667.1 | - | 9 | 832 | |||
BC030622.1 | - | 9 | 919 | 4818619 |