- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Gene Symbol: SOAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252512.1 | NP_001239441.1 | 2, 3 | 284 | ||
NM_003101.5 | NP_003092.4 | 3, 4 | 410 | |||
XM_011509911.1 | XP_011508213.1 | 3, 4 | 598 | |||
XM_011509912.1 | XP_011508214.1 | 3, 4 | 405 | |||
GenBank mRNA | AK026611.1 | - | 3, 4 | 322 | ||
AK300551.1 | - | 2, 3 | 208 | |||
AK316401.1 | - | 2, 3 | 113 | |||
BC028940.1 | - | 3, 4 | 242 | 4991365 | ||
DC344271.1 | - | 3, 4 | 410 | |||
L21934.2 | - | 1575 |