Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SOX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003106.3 | NP_003097.1 | 1 | 811 | ||
GenBank mRNA | AK312595.1 | - | 2 | 740 | ||
BC013923.2 | - | 1 | 449 | 2823424 | ||
KM822781.1 | - | 1 | 191 | |||
L07335.1 | - | 1 | 404 | |||
Z31560.1 | - | 1 | 386 |