Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGFBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000597.2 | NP_000588.2 | 1, 2 | 579 | ||
GenBank mRNA | A09809.1 | - | 1, 2 | 578 | ||
AB209509.1 | - | 1, 2 | 554 | |||
AL559513.3 | - | 1, 2 | 587 | |||
BC004312.1 | - | 1, 2 | 553 | 3627826 | ||
BC009902.1 | - | 1, 2 | 555 | 3509749 | ||
BC012769.1 | - | 1, 2 | 553 | 3689990 | ||
BC071967.1 | - | 1, 2 | 566 | 6650239 | ||
M35410.1 | - | 1, 2 | 522 | |||
X16302.1 | - | 1, 2 | 576 |