Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOXA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002141.4 | NP_002132.3 | 2 | 862 | ||
GenBank mRNA | BC126994.1 | - | 2 | 262 | ||
M74297.1 | - | 2 | 815 |