Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OAZ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301020.1 | NP_001287949.1 | 2 | 390 | ||
NM_004152.3 | NP_004143.1 | 2 | 396 | |||
GenBank mRNA | AF090094.1 | - | 1073 | |||
AK307221.1 | - | 2 | 362 | |||
BC010063.1 | - | 2 | 346 | |||
BI667301.1 | - | 2 | 367 | |||
BM423450.1 | - | 2 | 345 | |||
D78361.1 | - | 2 | 427 | |||
D87914.1 | - | 2 | 343 | |||
HY050868.1 | - | 2 | 396 | |||
U09202.1 | - | 2 | 328 |