Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APLNR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005161.4 | NP_005152.1 | 1 | 647 | ||
NR_027991.1 | 1 | 647 | ||||
GenBank mRNA | AK075252.1 | - | 1 | 395 | ||
AK097232.1 | - | 1 | 172 | |||
AK315542.1 | - | 1 | 444 | |||
AL548962.3 | - | 1 | 379 | |||
AL553350.3 | - | 1 | 648 | |||
BC032688.1 | - | 1 | 429 | 5241995 | ||
BX439288.2 | - | 1 | 633 | |||
X89271.1 | - | 1 | 507 |
Set Membership: |
![]() |